missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Tris Base, ULTROL™ Grade, Calbiochem™,
Useful in the pH range of 10.2 to 10.6
$204.00 - $2640.00
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Synonym | tris(Hydroxymethyl)aminomethane |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6483111KG
|
MilliporeSigma™
6483111KG |
1 kg |
Each for $204.00
|
|
|||||
6483115KG
|
MilliporeSigma™
6483115KG |
5 kg |
Each for $663.00
|
|
|||||
64831125KG
|
MilliporeSigma™
64831125KG |
25 kg |
Each for $2,640.00
|
|
|||||
Chemical Identifiers
77-86-1 | |
121.136 | |
tris(Hydroxymethyl)aminomethane | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
168°C to 172°C | |
219°C to 220°C | |
1 kg | |
C4H11NO3 | |
Soluble in water (800g/L at 20°C) | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
Tris Base |
77-86-1 | |
1.353g/cm3 | |
≥99.0% | |
tris(Hydroxymethyl)aminomethane | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.1 | |
by titration (dry basis) |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze).