missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™ Edge
Edge

DNase-, RNase- and Protease-Free.

Supplier:  Fisher BioReagents BP1525

 View more versions of this product

Catalog No. BP152-5


Edge
Explore available promotions

Description

Description

DNase-, RNase- and Protease-Free.

Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
Specifications

Specifications

0.05 AU max. at 280nm
168.5°C
219°C
10.4
5 kg
DNase-, RNase- and Protease-Free
Pass Test
C4H11NO3
Poly Pail
15, 9951
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
Molecular Biology
Tris Base
77-86-1
White
Crystalline Powder or Crystals
DNase free
2-Amino-2-(Hydroxymethyl)-1,3-Propanediol, Tris (hydroxymethyl) Aminomethane, Tris Base, THAM, Trimethylol Aminomethane, Trometamol
≥99.8 %
H2NC(CH2OH)3
Protease free
Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
C(C(CO)(CO)N)O
121.136
CHEBI:9754
≥99.8%

Chemical Identifiers

77-86-1
121.136
Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS Documents

Documents

Promotions

Promotions