missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(dibenzoylmethane)mono(phenanthroline)europium(III), Thermo Scientific Chemicals
As an organic red light emitter
$183.03 - $686.01
Chemical Identifiers
CAS | 17904-83-5 |
---|---|
Molecular Formula | C57H44EuN2O6 |
Molecular Weight (g/mol) | 1004.951 |
MDL Number | MFCD01321202 |
InChI Key | DYKOLWWJTALFFU-RWBKAWJDSA-N |
Synonym | eu dbm 3 phen, tris dibenzoylmethane mono phenanthroline europium iii, tris dibenzoylmethane phenanthroline europium iii, tris dibenzoylmethane mono 1,10-phenanthroline europium lll, europium iii tris 1,3-diphenyl-1,3-propanedionato mono 1,10-phenanthroline, 2e-3-bis 1e-3-oxo-1,3-diphenylprop-1-en-1-yl oxy europio oxy-1,3-diphenylprop-2-en-1-one; phen |
PubChem CID | 14205791 |
IUPAC Name | europium;(E)-3-hydroxy-1,3-diphenylprop-2-en-1-one;1,10-phenanthroline |
SMILES | C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu] |
Description
As an organic red light emitter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17904-83-5 | |
1004.951 | |
DYKOLWWJTALFFU-RWBKAWJDSA-N | |
14205791 | |
C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu] |
C57H44EuN2O6 | |
MFCD01321202 | |
eu dbm 3 phen, tris dibenzoylmethane mono phenanthroline europium iii, tris dibenzoylmethane phenanthroline europium iii, tris dibenzoylmethane mono 1,10-phenanthroline europium lll, europium iii tris 1,3-diphenyl-1,3-propanedionato mono 1,10-phenanthroline, 2e-3-bis 1e-3-oxo-1,3-diphenylprop-1-en-1-yl oxy europio oxy-1,3-diphenylprop-2-en-1-one; phen | |
europium;(E)-3-hydroxy-1,3-diphenylprop-2-en-1-one;1,10-phenanthroline |
Specifications
17904-83-5 | |
C57H44EuN2O6 | |
0.1 g | |
Soluble in dichloromethane, THF | |
C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu] | |
1004.951 | |
1001.93 | |
Tris (dibenzoylmethane)mono (phenanthroline)europium (III) |
172°C to 173°C | |
MFCD01321202 | |
eu dbm 3 phen, tris dibenzoylmethane mono phenanthroline europium iii, tris dibenzoylmethane phenanthroline europium iii, tris dibenzoylmethane mono 1,10-phenanthroline europium lll, europium iii tris 1,3-diphenyl-1,3-propanedionato mono 1,10-phenanthroline, 2e-3-bis 1e-3-oxo-1,3-diphenylprop-1-en-1-yl oxy europio oxy-1,3-diphenylprop-2-en-1-one; phen | |
DYKOLWWJTALFFU-RWBKAWJDSA-N | |
europium;(E)-3-hydroxy-1,3-diphenylprop-2-en-1-one;1,10-phenanthroline | |
14205791 | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only