missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
Tris-EDTA (TE) is routinely used for suspending nucleic acid samples.
Supplier: Fisher BioReagents BP2476500
Description
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
- Eliminates the hassle of dilution or waiting for powder to dissolve
- 1X solution contains 10 mM tris and 1 mM EDTA
- Filtered through a 5 micron filter and autoclaved
- Tested for the absence of DNase, RNase, and protease
- TE buffer is routinely used for resuspending nucleic acid samples
- Intended for laboratory research use only
NFPA:
- Health:1
- Flammability:0
- Instability:0
Specifications
Tris-EDTA | |
77-86-1,60-00-4,7647-01-0,7732-18-5 | |
7.4 to 8.1 | |
500 mL | |
Filtered through a 5-micron filter. | |
Protease free | |
Molecular Biology |
1X Solution, pH 7.4 | |
Colorless | |
Liquid | |
DNase free | |
Poly Bottle | |
TE |