Tris EDTA buffer, 1X, Molecular Biology Grade, Promega™
10mM Tris-HCl containing 1mM EDTA-Na2.
$56.60 - $209.27
Specifications
Description
- Quality tested and certified free of DNase, RNase and protease activity
Specifications
0.0 |