missing translation for 'onlineSavingsMsg'
Learn More

Tris-HCl, Molecular Grade, Promega™ DFS Item

Used in combination with Tris base for preparation of Tris-HCl buffers.

Supplier:  Promega Corporation H5121

 View more versions of this product

Catalog No. PR-H5121


Only null left
Explore available promotions

Description

Description

  • Quality tested and certified free of DNase, RNase and protease activity
Specifications

Specifications

Tris-HCl
100 g
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
Molecular Biology
1185-53-1
C4H12ClNO3
C(C(CO)(CO)N)O.Cl
157.594
157.56

Chemical Identifiers

1185-53-1
157.594
93573
C(C(CO)(CO)N)O.Cl
C4H12ClNO3
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15 to 30°C

SDS
Documents

Documents

Promotions

Promotions