Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
$67.25 - $458.97
Chemical Identifiers
| CAS | 1185-53-1 |
|---|---|
| Molecular Formula | C4H12ClNO3 |
| Molecular Weight (g/mol) | 157.594 |
| MDL Number | MFCD00012590 |
| InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
| Synonym | tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride |
| PubChem CID | 93573 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
| SMILES | C(C(CO)(CO)N)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1137918
|
Thermo Scientific Chemicals
A1137918 |
50 g |
Each for $67.25
|
|
|||||
|
AAA1137930
|
Thermo Scientific Chemicals
A1137930 |
250 g |
Each for $163.29
|
|
|||||
|
AAA113790B
|
Thermo Scientific Chemicals
A113790B |
1000 g |
Each for $458.97
|
|
|||||
Description
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
Chemical Identifiers
| 1185-53-1 | |
| 157.594 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 93573 | |
| C(C(CO)(CO)N)O.Cl |
| C4H12ClNO3 | |
| MFCD00012590 | |
| tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
| 1185-53-1 | |
| 3.5 to 5.0 | |
| (HOCH2)3CNH2·HCl | |
| 50 g | |
| Hygroscopic | |
| Soluble in water. | |
| C(C(CO)(CO)N)O.Cl | |
| 157.594 | |
| 157.6 | |
| Tris (hydroxymethyl)aminomethane hydrochloride |
| ∼152°C (decomposition) | |
| C4H12ClNO3 | |
| MFCD00012590 | |
| 3675235 | |
| tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
| 93573 | |
| ≥99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-684-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only