missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(pentafluorophenyl)borane, 95%, Thermo Scientific Chemicals

$277.98 - $989.21

Chemical Identifiers

CAS 1109-15-5
Molecular Formula C18BF15
Molecular Weight (g/mol) 511.98
MDL Number MFCD00269813
InChI Key OBAJXDYVZBHCGT-UHFFFAOYSA-N
Synonym tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
PubChem CID 582056
IUPAC Name tris(2,3,4,5,6-pentafluorophenyl)borane
SMILES FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC353130010
SDS View Documents
Thermo Scientific Chemicals
353130010
1 g Glass bottle
Each for $277.98
 
AC353130050
SDS View Documents
Thermo Scientific Chemicals
353130050
5 g Glass bottle
Each for $989.21
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1109-15-5
511.98
OBAJXDYVZBHCGT-UHFFFAOYSA-N
582056
FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
C18BF15
MFCD00269813
tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
tris(2,3,4,5,6-pentafluorophenyl)borane

Specifications

1109-15-5
94% min.
94% min
C18BF15
MFCD00269813
15, 9931
OBAJXDYVZBHCGT-UHFFFAOYSA-N
tris(2,3,4,5,6-pentafluorophenyl)borane
582056
95%
Tris(pentafluorophenyl)borane, 95%
126°C to 131°C
Authentic
Glass bottle
(C6F5)3B
1 g
tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
511.98
511.99
Powder
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only