Learn More
Tris(trimethylsilyloxy)ethylene, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 69097-20-7 |
---|---|
Molecular Formula | C11H28O3Si3 |
Molecular Weight (g/mol) | 292.60 |
MDL Number | MFCD00011641 |
InChI Key | FCZGHPGTZRTDNN-UHFFFAOYSA-N |
Synonym | tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene |
PubChem CID | 553067 |
IUPAC Name | 1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane |
SMILES | C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC216010010
|
Thermo Scientific Chemicals
216010010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
69097-20-7 | |
292.60 | |
FCZGHPGTZRTDNN-UHFFFAOYSA-N | |
553067 | |
C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C |
C11H28O3Si3 | |
MFCD00011641 | |
tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene | |
1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane |
Specifications
69097-20-7 | |
90.0°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4190 to 1.4220 | |
MFCD00011641 | |
08,523; 09,512 | |
tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene | |
FCZGHPGTZRTDNN-UHFFFAOYSA-N | |
1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane | |
553067 | |
95% |
0.8850g/mL | |
39°C | |
94% min. (GC) | |
C11H28O3Si3 | |
[(CH3)3SiO]2C=CHOSi(CH3)3 | |
1g | |
0.885 | |
Solubility in water: insoluble. | |
C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C | |
292.60 | |
292.59 | |
Tris(trimethylsilyloxy)ethylene |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 273-864-1
RUO â Research Use Only