missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris, Ultra Pure grade, 99.95% min., MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210313301

 View more versions of this product

Catalog No. ICN10313301


Explore available promotions

Description

Description

Specifications

Specifications

Tris
White
1 kg
C4H11NO3
2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Ultrapure
77-86-1
Crystalline Powder
>99.9%
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.1

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base
CHEBI:9754
C(C(CO)(CO)N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature, desiccate.

SDS
Documents

Documents

Promotions

Promotions