missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TRIS, USP grade, 99.0 to 101.0%, MP Biomedicals™
Tris is excellent biochemical and biological buffer for all applications where ultra high purity is required
Supplier: MP Biomedicals Inc 0219560580
Specifications
TRIS | |
White | |
100 g | |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 |
77-86-1 | |
Crystalline Powder | |
99.0 to 101.0% | |
Trometamol, Tris-[hydroxymethyl]aminomethane | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
USP |
Chemical Identifiers
77-86-1 | |
121.136 | |
Trometamol, Tris-[hydroxymethyl]aminomethane | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C). Desiccate.