missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trisodium N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetate Dihydrate 98.0+%, TCI America™
$51.90 - $192.05
Chemical Identifiers
CAS | 139-89-9 |
---|---|
Molecular Formula | C10H15N2Na3O7 |
Molecular Weight (g/mol) | 344.206 |
MDL Number | MFCD00043341 |
InChI Key | WHNXAQZPEBNFBC-UHFFFAOYSA-K |
Synonym | trisodium hedta, hamp-ol crystals, perma kleer 80, chemcolox 800, monaquest ica-120, versen-ol, chel dm 41, detarol trisodium salt, caswell no. 487c, hedta-na3 |
PubChem CID | 8772 |
IUPAC Name | trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate |
SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].[Na+].[Na+].[Na+] |
Chemical Identifiers
139-89-9 | |
344.206 | |
WHNXAQZPEBNFBC-UHFFFAOYSA-K | |
8772 | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].[Na+].[Na+].[Na+] |
C10H15N2Na3O7 | |
MFCD00043341 | |
trisodium hedta, hamp-ol crystals, perma kleer 80, chemcolox 800, monaquest ica-120, versen-ol, chel dm 41, detarol trisodium salt, caswell no. 487c, hedta-na3 | |
trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate |
Specifications
288°C | |
139-89-9 | |
25 g | |
MFCD00043341 | |
WHNXAQZPEBNFBC-UHFFFAOYSA-K | |
trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(2-hydroxyethyl)amino]acetate | |
8772 | |
≥98.0% (T) |
White | |
Crystalline Powder | |
C10H15N2Na3O7 | |
trisodium hedta, hamp-ol crystals, perma kleer 80, chemcolox 800, monaquest ica-120, versen-ol, chel dm 41, detarol trisodium salt, caswell no. 487c, hedta-na3 | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CCO)CC(=O)[O-].[Na+].[Na+].[Na+] | |
344.206 | |
344.21 | |
Trisodium N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetate Dihydrate |
Safety and Handling
EINECSNumber : (2)-1268
RTECSNumber : MB9188000
TSCA : Yes