missing translation for 'onlineSavingsMsg'
Learn More

Trisodium Naphthalene-1,3,6-trisulfonate Hydrate 65.0+%, TCI America™

$65.24 - $536.84

Chemical Identifiers

CAS 5182-30-9
Molecular Formula C10H5Na3O9S3
Molecular Weight (g/mol) 434.29
MDL Number MFCD00003989
InChI Key NJPKYOIXTSGVAN-UHFFFAOYSA-K
Synonym trisodium 1,3,6-naphthalenetrisulfonate, 1,3,6-naphthalenetrisulfonic acid, trisodium salt, naphthalene-1,3,6-trisulfonic acid trisodium salt, trisodium naphthalene-1,3,6-trisulfonate, sodium naphthalene-1,3,6-trisulfonate, trisodium 1,3,6-naphthalene trisulfonate, trisodium naphthalene-1,3,6-trisulphonate, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:3, 1,3,6-naphthalene trisulfonic acid trisodium salt, trisodium naphthalene trisulfonate
PubChem CID 78855
IUPAC Name trisodium naphthalene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N002125G
View Documents
TCI America
N002125G
25 g
Each for $65.24
Only null left
 
N0021100G
View Documents
TCI America
N0021100G
100 g
Each for $150.98
Only null left
 
N0021500G
View Documents
TCI America
N0021500G
500 g
Each for $536.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5182-30-9
434.29
NJPKYOIXTSGVAN-UHFFFAOYSA-K
78855
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O
C10H5Na3O9S3
MFCD00003989
trisodium 1,3,6-naphthalenetrisulfonate, 1,3,6-naphthalenetrisulfonic acid, trisodium salt, naphthalene-1,3,6-trisulfonic acid trisodium salt, trisodium naphthalene-1,3,6-trisulfonate, sodium naphthalene-1,3,6-trisulfonate, trisodium 1,3,6-naphthalene trisulfonate, trisodium naphthalene-1,3,6-trisulphonate, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:3, 1,3,6-naphthalene trisulfonic acid trisodium salt, trisodium naphthalene trisulfonate
trisodium naphthalene-1,3,6-trisulfonate

Specifications

5182-30-9
MFCD00003989
trisodium 1,3,6-naphthalenetrisulfonate, 1,3,6-naphthalenetrisulfonic acid, trisodium salt, naphthalene-1,3,6-trisulfonic acid trisodium salt, trisodium naphthalene-1,3,6-trisulfonate, sodium naphthalene-1,3,6-trisulfonate, trisodium 1,3,6-naphthalene trisulfonate, trisodium naphthalene-1,3,6-trisulphonate, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:3, 1,3,6-naphthalene trisulfonic acid trisodium salt, trisodium naphthalene trisulfonate
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O
434.29
434.29
Crystalline Powder
C10H5Na3O9S3
25 g
NJPKYOIXTSGVAN-UHFFFAOYSA-K
trisodium naphthalene-1,3,6-trisulfonate
78855
≥65.0% (HPLC)
Trisodium Naphthalene-1,3,6-trisulfonate Hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0471

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'