Learn More
Triton™ X-100 (Electrophoresis), Fisher BioReagents™
For solubilization and permeabilization of membranes
Chemical Identifiers
| CAS | 9002-93-1 |
|---|---|
| Molecular Formula | C16H26O2 |
| Molecular Weight (g/mol) | 250.38 |
| MDL Number | MFCD00132505 |
| InChI Key | JYCQQPHGFMYQCF-UHFFFAOYSA-N |
| Synonym | Polyethylene Glycol p-tert-Octylphenyl Ether |
| PubChem CID | 5590 |
| SMILES | CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
Description
This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.Chemical Identifiers
| C16H26O2 | |
| MFCD00132505 | |
| Polyethylene Glycol p-tert-Octylphenyl Ether | |
| CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
Specifications
| Undesignated | |
| Liquid | |
| 6°C | |
| 1.067g/cm³ | |
| Pass Test | |
| 4-(C8H17) C6H4 (OCH2CH2)n OH (n∽10) | |
| 15, 6850 | |
| Soluble in water | |
| CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 | |
| 250.38 | |
| 646.86 | |
| Triton™ X-100 |
| 9002-93-1 | |
| 270°C | |
| 63 to 69°C | |
| 6 to 8 | |
| C16H26O2 | |
| MFCD00132505 | |
| Polyethylene Glycol p-tert-Octylphenyl Ether | |
| JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
| 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol | |
| 5590 | |
| Electrophoresis |
Safety and Handling
WARNING!
Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artificial respiration with the aid of a pocket mask equipped with a one way valve or other proper respiratory medical device. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:1
Instability:1
ShelfLife : 2 years
Recommended Storage : RT