Hengar Boiling Granules Edge
Edge
Henger Granules effectively prevent superheating with its attendant dangerous explosive vaporization commonly known as bumping
Supplier:  Hengar 903800
 View more versions of this product
Catalog No. 02-215-521
Edge
Explore available promotions
Description
Description
  • Granules allow for a steady, copious stream of bubbles to escape without spasmodic splatter
  • Evaporation or distillation proceeds quietly and smoothly
  • Cannot contaminate the liquid
  • Reduced flask breakage
  • Minimized watching time
  • Quantity: 500g
Specifications
Specifications
Plain Micro Granules
Amphoteric Alundum
500 g