Troemner™ Individual Analytical Precision Weights, Class 1 with NVLAP Certificate
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Supplier:  Troemner™ 80781144
 View more versions of this product
Catalog No. 02-214-1JW
Explore available promotions
Description
Description
Designed so the specifications on surface finish, markings, construction, material density and magnetic properties get more restrictive as the weight tolerances get smaller in order to enable the weight to maintain its tolerance over time
  • Used in extremely stable environments
  • 1mg to 500mg are 1 piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are 2 piece construction with handling knob
  • Surfaces are polished to a perfect, mirror-like finish
Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.1mg to 0.01mg
Specifications
Specifications
Two-Piece Metric Analytical Weight
ASTM Class 1
200 g
200 g
Polycarbonate case
1
NVLAP, ISO9001
Stainless Steel
7.95g/cm3 at 20°C
Cylindrical with Handling Knob
Each