Troemner™ Individual Analytical Precision Weights, Class 1 with Traceable Certificate
Designed for your most demanding weighing applications
$324.71 - $4742.17
Specifications
Certifications/Compliance | Traceable certificate, ISO9001 |
---|---|
Classification | ASTM Class 1 |
Includes | Polycarbonate case |
Quantity | Each |
Catalog Number | Mfr. No. | Mass | Density | Material | Shape | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mass | Density | Material | Shape | Price | Quantity | ||
023181HT
|
Troemner™
30391598 |
500 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $954.66
|
|
||
02-215-100T
|
Troemner™
30391599 |
1 kg | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $1,845.94
|
|
||
02-214-1JT
|
Troemner™
30391596 |
200 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $744.34
|
|
||
02-319-1AT
|
Troemner™
30391595 |
100 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $467.03
|
|
||
023191BT
|
Troemner™
30391594 |
50 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $477.31
|
|
||
02-319-1CT
|
Troemner™
30391592 |
20 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $446.59
|
|
||
02-319-1DT
|
N/A | 10 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $497.38
|
|
||
02-319-1ET
|
N/A | 5 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $497.38
|
|
||
02-319-1FT
|
N/A | 2 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $477.07
|
|
||
023191GT
|
Troemner™
30391587 |
1 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $358.06
|
|
||
02-319-1HT
|
N/A | 500 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $395.76
|
|
||
023191JT
|
Troemner™
30391584 |
200 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $324.71
|
|
||
02-319-1KT
|
Troemner™
30391583 |
100 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $324.71
|
|
||
02-319-1LT
|
Troemner™
30391582 |
50 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $324.71
|
|
||
023191NT
|
Troemner™
30391580 |
20 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $324.71
|
|
||
02-319-1PT
|
N/A | 10 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $395.76
|
|
||
023191QT
|
Troemner™
30391578 |
5 mg | 7.95g/cm3 at 20°C | Stainless Steel | Flat with one side turned up |
Each for $324.71
|
|
||
023191RT
|
Troemner™
30391576 |
2 mg | 2.7g/cm3 at 20°C | Aluminum | Flat with one side turned up |
Each for $324.71
|
|
||
02-319-1TT
|
N/A | 1 mg | 2.7g/cm3 at 20°C | Aluminum | Flat with one side turned up |
Each for $384.21
|
|
||
02-214-077
|
Troemner™
30391589 |
3 g | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $456.79
|
|
||
02-225-539
|
N/A | 5 kg | 7.95g/cm3 at 20°C | Stainless Steel | Cylindrical with Handling Knob |
Each for $4,742.17
|
|
||
Description
Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
- Precision Analytical Individual Weights are made from the finest materials; manufactured in sizes ranging from 1mg through 50kg
- Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
- Fractional weights 3mg and below are made of aluminum
- Weights 1mg to 2kg are supplied in a high-quality polycarbonate case
- Used as reference standards as well as calibrating Class I balances as specified in OIML R 76
- Used to calibrate Class 1 weights
- Calibrate certain Class I and Class II balances and scales as specified in OIML R 76
- Can also be used to calibrate Class 2 weights
Specifications
Traceable certificate, ISO9001 | |
ASTM Class 1 | |
Polycarbonate case | |
Each |