Troemner™ Individual Analytical Precision Weights, Class 3 with NVLAP Certificate
Designed for your most demanding weighing applications
$320.26 - $4881.66
Specifications
Certifications/Compliance | NVLAP, ISO9001 |
---|---|
Classification | ASTM Class 3 |
Includes | Polycarbonate case |
Quantity | Each |
Catalog Number | Mfr. No. | Mass | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mass | Price | Quantity | |||||
02-214-128
|
N/A | 2 kg |
Each for $3,223.72
|
|
|||||
02-214-127
|
Troemner™
80858170 |
200 g |
Each for $709.47
|
|
|||||
02-215-30CC
|
Troemner™
80858489 |
50 mg |
Each for $320.26
|
|
|||||
02-225-504
|
N/A | 20 g |
Each for $489.48
|
|
|||||
02-225-510
|
N/A | 2 g |
Each for $452.66
|
|
|||||
02-225-512
|
N/A | 500 mg |
Each for $378.97
|
|
|||||
02-225-522
|
N/A | 20 mg |
Each for $378.97
|
|
|||||
02-225-524
|
N/A | 10 mg |
Each for $378.97
|
|
|||||
02-225-527
|
N/A | 5 mg |
Each for $378.97
|
|
|||||
02-225-530
|
N/A | 2 mg |
Each for $378.97
|
|
|||||
02-225-532
|
N/A | 1 mg |
Each for $378.97
|
|
|||||
02-225-545
|
N/A | 500 g |
Each for $1,110.55
|
|
|||||
02-225-546
|
N/A | 1 kg |
Each for $2,107.93
|
|
|||||
02-225-547
|
N/A | 5 kg |
Each for $4,881.66
|
|
|||||
Description
Low magnetic susceptibility, uniform consistency and high resistance to corrosion. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1g to 0.01g
- Weights 1mg through 2kg are supplied in a high-quality polycarbonate case
Specifications
NVLAP, ISO9001 | |
ASTM Class 3 | |
Polycarbonate case | |
Each |