missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ Individual Analytical Precision Weights, Class 4 with NVLAP Certificate
Designed for your most demanding weighing applications
$422.56 - $3958.74
Specifications
Certifications/Compliance | NVLAP, ISO9001 |
---|---|
Classification | ASTM Class 4 |
Material | Stainless Steel |
Density | 7.95g/cm3 at 20°C |
Includes | Polycarbonate case |
Catalog Number | Mfr. No. | Mass | Shape | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mass | Shape | Price | Quantity | ||||
02-214-134
|
Troemner™
30391323 |
5 kg | Cylindrical with Handling Knob |
Each for $3,958.74
|
|
||||
02-214-132
|
N/A | 500 g | Cylindrical with Handling Knob |
Each for $1,044.76
|
|
||||
02-214-133
|
Troemner™
30391321 |
2 kg | Cylindrical with Handling Knob |
Each for $2,628.76
|
|
||||
02-225-541
|
N/A | 200 g | Cylindrical with Handling Knob |
Each for $757.90
|
|
||||
02-225-542
|
N/A | 1 kg | Cylindrical with Handling Knob |
Each for $1,952.66
|
|
||||
02-100-153
|
Troemner™
80850287 |
100 g | Cylindrical with Handling Knob |
Each for $422.56
|
|
||||
Description
Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
- Precision Analytical Individual Weights are made from the finest materials; manufactured in sizes ranging from 1mg through 50kg
- Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
- Fractional weights 3mg and below are made of aluminum
- Weights 1mg to 2kg are supplied in a high-quality polycarbonate case
- Reference standards as well as calibrating Class I balances as specified in OIML R 76
- Calibration of semi-analytical balances and for student use
Order Info
Weights supplied in a rugged, precision-fit, high quality polycarbonate case.
Specifications
NVLAP, ISO9001 | |
Stainless Steel | |
Polycarbonate case |
ASTM Class 4 | |
7.95g/cm3 at 20°C | |
Each |