Troemner™ Individual Analytical Precision Weights, Class 4
Designed for your most demanding weighing applications
 View more versions of this product
Catalog No. 02-214-042
Explore available promotions
Description
Description
Low magnetic susceptibility, uniform consistency and high resistance to corrosion. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • For calibration of semi-analytical balances and for student use.
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
Specifications
Specifications
One Piece Metric Analytical Sheet Weight
Aluminum
2.7g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
ASTM Class 4
1 mg
1 mg
Polycarbonate case
1