Troemner™ NIST™ Class F Stainless-Steel Avoirdupois Weights
Meet or exceed tolerances established for Class F in NIST Handbook 105-1
Description
- Available in avoirdupois and metric sets
- Can be used in the field and for onsite servicing applications
- Packed in sturdy polypropylene case with handle
Specifications
Small Avoirdupois Test Weight Set | |
NIST Class F 105-1 | |
7.85g/cm3 at 20°C | |
Cylindrical weight: 5 lb., 2 x 2 lb., 1 lb., 8 oz., 4 oz., 2 oz., 1 oz., 0.5 oz., 0.25 oz., 0.12 oz., 0.06 oz., sheet weight: 2 x 0.03 oz., Plastic case | |
Multiple |
Statement of accuracy, ISO9001 | |
Stainless Steel | |
1/3 oz. to 5 lb. | |
14/Set |