Learn More
Troemner™ Slotted Aluminium Weight Hanger
For use with slotted Aluminium sets and individual weights.
Supplier:  Troemner™ 30390750
Catalog No. S97603A
Only null left
Explore available promotions
Description
Description
For use with slotted Aluminium sets and individual weights.
  • Weight hanger is calibrated to 50g
Specifications
Specifications
Weight Hanger
Stainless Steel
50 g
Each
ANSI/ASTM Class 7
7.85g/cm3 at 20°C
62 mg
Product Suggestions
Product Suggestions