Troemner™ Talboys™ Heavy-Duty Lab-Lifts
With over-sized, sure-grip adjustment knobs that provide extra leverage for easy height adjustments
Supplier: Troemner™ 30400002
Description
- Heavy duty 300 series stainless steel construction
- Chemical resistant
- Autoclavable
- Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
- Suited for use in fume hoods or bench tops
- Can withstand extreme environments and high load applications
- Available in different sizes
Specifications
Stainless Steel | |
9.75 to 3 in. | |
35 lb. | |
Autoclavable |
15.2 to 15.2 cm | |
6 x 6 in. | |
Talboys™ Heavy Duty Lab Lifts |