Troemner™ Talboys™ Heavy-Duty Lab-Lifts
With over-sized, sure-grip adjustment knobs that provide extra leverage for easy height adjustments
Supplier:  Troemner™ 30400005
 View more versions of this product
Catalog No. 02-214-206
Explore available promotions
Description
Description
  • Heavy duty 300 series stainless steel construction
  • Chemical resistant
  • Autoclavable
  • Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
  • Suited for use in fume hoods or bench tops
  • Can withstand extreme environments and high load applications
  • Available in different sizes
Specifications
Specifications
Stainless Steel
19.5 to 4 in.
150 lb.
Autoclavable
30.4 to 30.4 cm
12 x 12 in.
Talboys™ Heavy Duty Lab Lifts
Videos
SDS
Documents
Documents
Accessories