Troemner™ Talboys™ Heavy-Duty Lab-Lifts
With over-sized, sure-grip adjustment knobs that provide extra leverage for easy height adjustments
Supplier:  Troemner™ 30400006
 View more versions of this product
Catalog No. 02-214-207
Explore available promotions
Description
Description
  • Heavy duty 300 series stainless steel construction
  • Chemical resistant
  • Autoclavable
  • Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
  • Suited for use in fume hoods or bench tops
  • Can withstand extreme environments and high load applications
  • Available in different sizes
Specifications
Specifications
Stainless Steel
19.5 to 4 in.
180 lb.
Autoclavable
40.6 to 40.6 cm
16 x 16 in. (Deck)
Talboys™ Heavy Duty Lab Lifts
Videos
SDS
Documents
Documents
Accessories