Troemner™ Talboys™ Labjaws™ Lab-Frame Kits and Rods
Available in aluminum, fiberglass and stainless-steel and are corrosion resistant
$29.62 - $6306.32
Specifications
For Use With (Equipment) Troemner Talboys
View More Specs
Products 30
Catalog Number Mfr. No. Material Product Type Length (English) Includes Price Quantity  
Catalog Number Mfr. No. Material Product Type Length (English) Includes Price Quantity  
02-214-154
View Documents
Troemner™
30392280
Aluminum Lab Frame Rod, Chamfered Ends 72 in.
Each for $201.33
02-214-155
View Documents
Troemner™
30392274
Aluminum Lab Frame Rod 12 in.
Each for $68.73
02-214-152
View Documents
Troemner™
30392275
Aluminum Lab Frame Rod, Chamfered Ends 18 in.
Each for $74.03
02-214-153
View Documents
Troemner™
30392277
Aluminum Lab Frame Rod, Chamfered Ends 36 in.
Each for $97.71
02-214-156
View Documents
Troemner™
30392281
Aluminum Lab Frame Rod, Chamfered Ends 96 in.
Each for $253.18
02-214-158
View Documents
Troemner™
30392305
Aluminum Lab Frame Eight 2 in. rods, two 18 in. rods, seven 24 in. rods, four 48 in. rods, 35 S-connectors, 4 rod end connectors, 4 lab frame feet
Each for $2,406.79
02-300-225
View Documents
Troemner™
30392263
Nickel Plated Zinc Lab Frame Foot 1.9cm mounting screws
Each for $103.24
02-300-227
View Documents
Troemner™
30392276
Aluminum Lab Frame Rod, Chamfered Ends 24 in.
Each for $91.79
02-300-228
View Documents
Troemner™
30392278
Aluminum Lab Frame Rod, Chamfered Ends 48 in.
Each for $153.97
02-300-230
View Documents
Troemner™
30392298
Stainless Steel Lab Frame Rod, Chamfered Ends 24 in.
Each for $155.58
02-300-229
View Documents
Troemner™
30392297
Stainless Steel Lab Frame Rod 18 in.
Each for $125.06
02-300-232
View Documents
Troemner™
30392304
Aluminum Lab Frame Kit Eight 2 in. rods, two 18 in. rods, eight 24 in. rods, 18 S-connectors, 4 rod end connectors, 4 lab frame feet
Each for $1,856.35
02-300-231
View Documents
Troemner™
30392299
Stainless Steel Lab Frame Rod 36 in.
Each for $210.48
02-300-234
View Documents
Troemner™
30392307
Aluminum Lab Frame Six 2 in. rods, six 12 in. rods, two 24 in. rods, seven 48 in. rods, five 72 in. rods, 45 S-connectors, 4 rod end connectors, 8 lab frame feet
Each for $4,982.88
02-300-245
View Documents
Troemner™
30392272
Aluminum Lab Frame Rod, Chamfered Ends 2 in.
Each for $41.44
02-216-270
View Documents
Troemner™
30392273
Aluminum Lab Frame Rod, Chamfered Ends 6 in.
Each for $34.28
02-216-269
View Documents
Troemner™
30392271
Aluminum Lab Frame Rod 1.61 in.
Each for $29.62
02-216-271
View Documents
Troemner™
30392279
Aluminum Lab Frame Rod, Chamfered Ends 60 in.
Each for $177.65
02-216-286
View Documents
Troemner™
30392296
Stainless Steel Lab Frame Rod, Chamfered Ends 12 in.
Each for $94.58
02-216-285
View Documents
Troemner™
30392295
Stainless Steel Lab Frame Rod, Chamfered Ends 6 in.
Each for $56.26
02-216-284
View Documents
Troemner™
30392294
Stainless Steel Lab Frame Rod, Chamfered Ends 2 in.
Each for $44.41
02-216-283
View Documents
Troemner™
30392293
Stainless Steel Lab Frame Rod, Chamfered Ends 1.61 in.
Each for $35.53
02-216-287
View Documents
Troemner™
30392300
Stainless Steel Lab Frame Rod 48 in.
Each for $260.53
02-216-288
View Documents
Troemner™
30392301
Stainless Steel Lab Frame Rod 60 in.
Each for $304.94
02-216-289
View Documents
Troemner™
30392302
Stainless Steel Lab Frame Rod, Chamfered Ends 72 in.
Each for $364.18
02-216-290
View Documents
Troemner™
30392303
Stainless Steel Lab Frame Rod, Chamfered Ends 96 in.
Each for $473.71
02-216-310
View Documents
Troemner™
30392324
Stainless Steel Lab Frame
Each for $3,244.81
02-216-311
View Documents
Troemner™
30392325
Stainless Steel Lab Frame
Each for $6,306.32
02-216-309
View Documents
Troemner™
30392323
Stainless Steel Lab Frame Kit Eight 2 in. rods, two 18 in. rods, eight 24 in. rods, 18 S-connectors, 4 rod end connectors, 4 lab frame feet
Each for $3,162.03
02-216-312
View Documents
Troemner™
30392326
Stainless Steel Lab Frame Kit Six 2 in. rods, six 18 in. rods, two 24 in. rods, seven 48 in. rods, five 72 in. rods, 45 S-connectors, 4 rod end connectors, 8 lab frame feet
Each for $5,122.93
Description
Description
Lab-Frame Kits
  • Available in heavy-duty, extra large, large, medium, and small
  • Heavy-duty lab frame
    • Designed for intense work like pilot plant setups
    • Freestanding unit
    • Dimensions: 48 x 72 in. (121.9 x 182.9cm)
    • Outside frame is made of lightweight steel
    • Interior lattice is comprised of lab frame rods and horizontal channel connectors
    • One end accommodates the rod, other end fits into channel and locks tightly with a bolt
    • On loosening bolt, connector slides easily along the channel track allowing for simple lattice adjustment
    • Apparatus is supported by sturdy, counter-weighted cast iron feet that can be bolted to floor
    Extra-large lab frame
    • Equipped for horizontal or vertical use
    • Measures 48 x 72 in. (121.9 x 182.9cm)
    Large lab frame
    • Accommodates complex glassware setups
    • Measures 48 x 48 in. (121.9 x 121.9cm)
    • Base is 18 in. (46cm) wide and can be mounted on floor for additional stability
    Medium lab frame
    • Convenient distillation or general setup
    • Frame can be assembled in two ways: 24H x 48 in. L (61 x 121.9cm) horizontally or 48H x 24 in. L (121.9 x 61cm) vertically
    • Small lab frame
      • Ideal for small set-ups and limited spaces
      • Measures 24 x 24 in. (61 x 61cm)
      • Can be permanently mounted to the bench top
      Lab-frame rods
      • Chamfered ends for smooth fit
      • Available in standard lengths from 1.61 in. (4.1cm) to 96 in. (243.8cm)
      • Diameter: 0.51 in. (1.3cm)
      Lab-frame foot
      • Used for mounting lattice permanently to floors, walls or bench tops
      • Equipped with three screws
      • Low profile design
      • Maximum rod size: 0.51 in. (1.3cm)
    Specifications
    Specifications
    Troemner Talboys
    Videos
    SDS
    missing translation for 'documents'
    missing translation for 'documents'