missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trolox(R), 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 218940050

 View more versions of this product

Catalog No. AC218940050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trolox*
53188-07-1
Beige to White
97%
C14H18O4
5 g
trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm
GLEVLJDDWXEYCO-UHFFFAOYNA-N
6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid
40634
250.29
Crystalline Powder
97%
187.0°C to 194.0°C
Authentic
Glass bottle
MFCD00006846
18, V,7, 335
Solubility in water: slightly soluble. Other solubilities: soluble in methanol, ethanol
CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O
250.29
CHEBI:82625
97%

Chemical Identifiers

53188-07-1
250.29
GLEVLJDDWXEYCO-UHFFFAOYNA-N
40634
6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydrochromene-2-carboxylic acid
C14H18O4
MFCD00006846
trolox, 6-hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid, trolox c, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-2h-1-benzopyran-2-carboxylic acid, 6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl, +/--6-hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro-6-hydroxy-2,5,7,8-tetramethyl-, +-, trolox™, trolox tm
CHEBI:82625
CC1=C(C)C2=C(CCC(C)(O2)C(O)=O)C(C)=C1O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-422-8

RTECSNumber : DJ2273000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only