missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tropine, 98%, Thermo Scientific Chemicals

$85.84 - $277.58

Chemical Identifiers

CAS 120-29-6
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00005551
InChI Key CYHOMWAPJJPNMW-RNFRBKRXSA-N
Synonym tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
PubChem CID 6101956
IUPAC Name (1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES CN1C2CCC1CC(C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0233209
View Documents
Thermo Scientific Chemicals
L0233209
10 g
Each for $85.84
 
AAL0233218
View Documents
Thermo Scientific Chemicals
L0233218
50 g
Each for $277.58
 
Description

Description

Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tropine as an oxidative product of Tropane, used to synthesize alkaloid derivatives. Benzatropine and etybenzatropine are derivatives of tropine. It is also a building block of atropine, an anticholinergic drug prototypical of the muscarinic antagonist class.

Solubility
Soluble in water. (100 g/L) at 20°C.

Notes
Hygroscopic. Store at 4°C. Incomaptible with Strong acids and oxidizing agents.
Specifications

Chemical Identifiers

120-29-6
141.214
CYHOMWAPJJPNMW-RNFRBKRXSA-N
6101956
CN1C2CCC1CC(C2)O
C8H15NO
MFCD00005551
tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol

Specifications

120-29-6
1.04
232°C to 234°C
C8H15NO
10 g
Hygroscopic
tropine, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-methyl-8-azabicyclo 3.2.1 octan-3-ol, spectrum3_001840, 1r,5r-8-methyl-8-azabicyclo 3.2.1 octan-3beta-ol
CYHOMWAPJJPNMW-RNFRBKRXSA-N
(1R,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
6101956
98%
61°C to 65°C (dried)
12
Amine-like
MFCD00005551
80188
14,9781
Soluble in water. (100g/L) at 20°C.
CN1C2CCC1CC(C2)O
141.214
141.22
Tropine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 204-384-2

RTECSNumber : YM3875000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only