missing translation for 'onlineSavingsMsg'
Learn More

Trospium Chloride 98.0+%, TCI America™

$126.76 - $380.27

Chemical Identifiers

CAS 10405-02-4
Molecular Formula C25H30ClNO3
Molecular Weight (g/mol) 427.969
MDL Number MFCD00865254
InChI Key RVCSYOQWLPPAOA-VROPFNGYSA-M
PubChem CID 73415998
IUPAC Name [(1S,5S)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride
SMILES C1CC[N+]2(C1)C3CCC2CC(C3)OC(=O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T3305200MG
View Documents
TCI America
T3305200MG
200 mg
Each for $126.76
Only null left
 
T33051G
View Documents
TCI America
T33051G
1 g
Each for $380.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10405-02-4
427.969
RVCSYOQWLPPAOA-VROPFNGYSA-M
[(1S,5S)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride
C25H30ClNO3
MFCD00865254
73415998
C1CC[N+]2(C1)C3CCC2CC(C3)OC(=O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O.[Cl-]

Specifications

10405-02-4
White
MFCD00865254
RVCSYOQWLPPAOA-VROPFNGYSA-M
[(1S,5S)-spiro[8-azoniabicyclo[3.2.1]octane-8,1'-azolidin-1-ium]-3-yl] 2-hydroxy-2,2-diphenylacetate;chloride
73415998
≥98.0% (HPLC,N)
Trospium Chloride
262°C
C25H30ClNO3
200 mg
C1CC[N+]2(C1)C3CCC2CC(C3)OC(=O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O.[Cl-]
427.969
427.97
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : WH1424000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'