Learn More
Thermo Scientific™ TSG Series General Purpose Laboratory Freezers
Thermo Scientific™ TSG Series general purpose freezers are designed to meet the general storage needs of research, clinical and industrial laboratories.
$16387.33 - $22614.51
Specifications
Temperature Range | -12°C to -30°C |
---|---|
Temperature (Setpoint) | -23C° |
Door Style | Solid |
Door Material | Stainless Steel |
Cabinet Material | Stainless Steel |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Price | Quantity | ||||
TSG25FSSA
|
Thermo Scientific™
TSG25FSSA |
25 cu. ft. | 700 L |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
TSG49FSSA
|
Thermo Scientific™
TSG49FSSA |
49 cu. ft. | 1387 L |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Thermo Scientific™ TSG Series general purpose laboratory auto-defrost freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
Utilizing natural refrigerants in accordance with the U.S. Environmental Protection Agency (EPA) Significant New Alternative Policy (SNAP) and energy efficient systems that enable ENERGY STAR certification, the TSG Series improves scientific and healthcare sustainability.
The TSG Series general purpose laboratory freezers provide a reliable, quiet and efficient work environment.
Enabling lab efficiency and compliance- Designed to safely store reagents, media, proteins, and samples used daily in the lab which require great performance
- Adaptive defrost with time and temperature guided cycle provides safe storage temperatures and allows for continuous use without the need for defrost
- Easy to use digital controller with setpoint security and alarms (temperature, door ajar, and power failure)
- Tested and certified to conform with the design and safety standards of UL 471
- Easy to clean stainless steel surfaces and epoxy coated shelves
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the freezer without distraction
- Self-closing door with 90° stop to assist with inventory loads
- Standard door locks provide comfort and assurance
- US EPA ENERGY STAR Certified
- Natural refrigerants with near 0 GWP and ODP make the TSG Series compliant with US EPA’s Significant New Alternatives Policy (SNAP)
- Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
- Built in remote alarm terminals
- 24 Months parts and labor, 60-month compressor parts warranty (varies by country, please contact us for details)
Specifications
-12°C to -30°C | |
Solid | |
Stainless Steel | |
4 | |
45 kg | |
Digital Micrprocessor | |
2 years parts and labor, plus 3-year compressor warranty | |
Key Lock | |
115V 60Hz | |
R290 |
-23C° | |
Stainless Steel | |
Stainless Steel | |
100 lb. | |
ENERGYSTAR, Prop 65, cULus | |
1 Rear Access Port | |
Automatic | |
NEMA 5-15 P | |
60 Hz |