Learn More
Thermo Scientific™ TSG Series Undercounter Refrigerators
Benefit from innovative refrigeration solutions for healthcare and research needs
$6390.98 - $7231.11
Specifications
Capacity (English) | 5.5 cu. ft. |
---|---|
Capacity (Metric) | 156 L |
Temperature Range | 2°C to 9°C |
No. of Doors | 1 |
Hinge Side | Right |
Catalog Number | Mfr. No. | Plug Type | Electrical Requirements | Door Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Plug Type | Electrical Requirements | Door Style | Price | Quantity | |||
TSG505GA
|
Thermo Scientific™
TSG505GA |
NEMA 5-15 P, More Global Types Available | 100 - 240 V 50-60 Hz | Glass |
Each for $7,231.11
|
|
|||
TSG505SA
|
Thermo Scientific™
TSG505SA |
NEMA 5-15 P, More Global Types Available | 100 - 240 V 50-60 Hz | Solid |
Each for $6,390.98
|
|
|||
Description
Thermo Scientific™ TSG Series, powered by solid state refrigeration technology, has been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where solid performance is required.
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSG Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
- Designed to safely store media, reagents, pharmaceuticals, vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 2°C to 8°C
- Full alarm set, including hi/lo temperature, door ajar and power fail to support requirements for medical storage
- Remote alarm contacts on the undercounter refrigerator, supporting requirements of research for monitoring
- Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations
- Tested to GMP Clean Room Class A / ISO 7 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
- Tested and certified to conform with the design and safety standards of UL 61010 and CE Directive 2011/65/EU and 2015/863
- ADA compliant
- Ergonomically designed door handle
- Touchscreen controller with password protection is angled to help prevent neck strain
- Whisper quiet with a sound level of 35 dBA, enabling end users to work alongside the refrigerator without distraction
- Bright, easy-to-adjust interior LED lighting
- Self-closing door with stop to assist with inventory loads
- Universal voltage (100-240v, 50/60hz) allows placement throughout the facility and globally just by connecting the region-specific power cord.
- US EPA ENERGY STAR Certified with up to 70% less energy usage than conventional refrigerant models
- Natural, CO2 refrigerant and green foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
- Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
- Choice of reversible glass or solid door
- Extra and Alternate wire baskets and shelves
- Specialty, regional, and country specific plugs
- Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
- Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
Specifications
5.5 cu. ft. | |
2°C to 9°C | |
Right | |
Powder-coated CRS | |
Undercounter | |
Remote Alarm Contacts | |
684 mm | |
100 - 240 V 50-60 Hz |
156 L | |
1 | |
3 | |
CE, UL | |
No | |
26.9 in. (door closed) | |
100/240 V | |
Energy efficient |