missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ TSH Explosion Proof Combination Refrigerator/Freezer

Prevent the triggering of an explosion with this specially crafted, combination refrigerator-freezer

Supplier:  Thermo Scientific™ TSH07CESA

Catalog No. TSH07CESA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Prevent the triggering of an explosion inside or outside of the unit with Thermo Scientific™ Explosion-Proof Refrigerator/Freezers. Suitable for use in Class I, Division I, Group C and D hazaRDO™us environments, this device meets OSHA and National Fire Protection Association safety requirements.

  • Green, natural HC refrigerants
  • Thermostat and compressor components are isolated within a vapor-proof enclosure
  • Electrical connections are hardwired to prevent sparking and accidental disconnection
  • Magnetic vinyl gaskets for positive door seal CFC-free insulation and refrigerant
  • Manual defrost
  • Warranty varies by country, please contact us for details
Specifications

Specifications

Explosion-Proof Freezer/Refrigerator
201 L
Adjustable
Hinged
3 Adjustable Shelves, 1 Drawer (Refrigerator); 1 Shelf (Freezer)
Enameled Steel
ABS Plastic
Temperature Display - None
Mechanical Dial Thermostat
13 Months
None
Explosion Proof Combination Refrigerator/Freezer
Ref: 17 x 17.5 x 36.5 in. (43.2 x 44.5 x 92.7 cm)
Frz: 16 x 15.5 x 15.5 in. (40.6 x 39.4 x 26.4 cm)
115 V
60 Hz
45.8 kg
7.1 cu. ft.
1°C to 12°C
R600a
2 (2 compartments)
3
Cyclopentane
cULus
White
Indoor Cold Storage Of Materials In Class I, Division I Group C and D, Class I Zone 1 Group IIB Environments
Automatic (Refrigerator), Manual (Freezer)
12 in. at the top, 4 in. at the rear and 4 in. on each side
27.5 x 21.5 x 56.63 in. (69.8 x 54.6 x 143.8cm)
No Plug, Hardwired
115 V/60 Hz
101 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions