Learn More
Thermo Scientific™ TSX Series High-Performance Undercounter Lab Refrigerators
For a better environment inside and out - Introducing our new undercounter lab refrigerator.
$12451.56 - $12451.56
Specifications
Capacity (English) | 5.5 cu. ft. |
---|---|
Capacity (Metric) | 156 L |
No. of Doors | 1 |
Shelves | 3 |
Certifications/Compliance | CE, REACH, RoHS, UL |
Catalog Number | Mfr. No. | Door Style | Electrical Requirements | Plug Type | Price | Qty | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Door Style | Electrical Requirements | Plug Type | Price | Qty | |||
TSX505SA
|
Thermo Scientific™
TSX505SA |
Single Solid | 100 - 240 V 50-60 Hz | NEMA 5-15 P, More Global Types Available |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C. The Series' V-drive is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
Unique to the TSX505, our dual refrigeration solution operates through the action of two synchronized solid-state heat pumps. The two solid-state heat pumps work to cool a combination of cold wall and forced air to help ensure that conditions are optimal for the most sensitive applications
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
- Designed to safely store media, reagents, pharmaceuticals, vaccines, chemotherapy and other medical-grade storage requiring a temperature range of 2°C to 8°C
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Redundant cooling offers additional peace of mind for your most precious samples
- Automated electronic passcode locking standard
- Datalogging with USB download functionality
- Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations
- Tested to GMP Clean Room Class A / ISO 7 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
- Tested and certified to conform with the design and safety standards of UL 61010 and CE Directive 2011/65/EU and 2015/863
- ADA compliant
- Ergonomically designed door handle
- Touchscreen controller with password protection is angled to help prevent neck strain
- Whisper quiet with a sound level of 35 dBA, enabling end users to work alongside the refrigerator without distraction
- Bright, easy-to-adjust interior LED lighting
- Self-closing door with stop to assist with inventory loads
- Universal voltage (100-240v, 50/60hz) allows placement throughout the facility and globally just by connecting the region-specific power cord.
- US EPA ENERGY STAR Certified with up to 70% less energy usage than conventional refrigerant models
- Natural, CO2 and Water refrigerants and green foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
- Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems
- Touchscreen controller with password protection
- Choice of reversible glass or solid door
- Extra and Alternate wire baskets and shelves
- Specialty, regional, and country specific plugs
- Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate
- Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs
Specifications
5.5 cu. ft. | |
1 | |
CE, REACH, RoHS, UL | |
Alarm | |
20.5 in. | |
52 cm | |
23 in. | |
23.6 in. | |
60 cm | |
100 - 240 V 50-60 Hz |
156 L | |
3 | |
Automatic | |
26 in. | |
66 cm | |
31.8 in. | |
58.4 cm | |
19.5 in. | |
10 A Max / 3 A Steady State | |
50/60 Hz |