Learn More
Labcon North America TUBE 13X100 PS STERILE CS/1000 DFS Item
Supplier:  Labcon North America 34253550009
This item has a minimum order quantity of 3 per supplier requirements. Tube, Culture; Labcon; SuperClear; Polystyrene; Disposable; With extra thick wall; Without cap; Non-pyrogenic; Sterile; Size: 13 x 100mm; Capacity: 8mL; 125/bag
Catalog No. 50101531
Only null left
Explore available promotions