Eppendorf™ Tube Adapters for Eppendorf™ Model 5702 Round Buckets
Fit in round bucket 05-400-320 to accept tubes from 1.1 to 85mL
$406.10 - $779.03
Specifications
For Use With (Equipment) | A-4-38 swing bucket rotor (round bucket) for 5702/5702R/5702RH centrifuge |
---|
Catalog Number | Mfr. No. | No. of Places | Capacity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Places | Capacity | Price | Quantity | ||||
05-400-325
|
Eppendorf™
022639269 |
4 | 4 to 10 mL |
Pack of 2 Each for $730.25
|
|
||||
05-400-322
|
Eppendorf™
022639129 |
4 | 9 to 15 mL |
Pack of 2 Each for $731.49
|
|
||||
05-400-326
|
Eppendorf™
022639285 |
5 | 1.1 to 1.4 mL |
Pack of 2 Each for $779.03
|
|
||||
05-400-321
|
Eppendorf™
022639200 |
2 | 15 mL Conical Tubes |
Pack of 2 Each for $731.49
|
|
||||
05-400-324
|
Eppendorf™
022639226 |
1 | 50 mL Conical Tubes |
Pack of 2 Each for $406.10
|
|
||||
05-400-323
|
Eppendorf™
022639188 |
1 | 15 mL Conical Tubes |
Pack of 2 Each for $406.10
|
|
||||
05-400-317
|
Eppendorf™
022639242 |
4 | 2 to 7 mL |
Pack of 2 Each for $731.49
|
|
||||
Description
Adapters are sold in packs of two. Be sure to order proper number to fully equip your rotor.
Specifications
A-4-38 swing bucket rotor (round bucket) for 5702/5702R/5702RH centrifuge |