missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten foil, 2.0mm (0.08in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Form: approx. 24g/25x50mm

$1363.37 - $1363.37

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13980FY
View Documents
Thermo Scientific Chemicals
013980FY
50 x 100 mm
Each for $1,363.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]

Specifications

Silver to Gray
Silver Gray foil
99.95% (Metals basis)
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Tungsten
7440-33-7
19.3g/cm3
W
50 x 100 mm
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Foil
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only