missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten hexacarbonyl, 99%, Thermo Scientific Chemicals

Tungsten hexacarbonyl, 99%, C6O6W, CAS Number-14040-11-0, 250g, 175.0 deg.C, Yellow to Beige, 237-880-2, 98.5% min., 06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51, 351.91, GHS Signal Word: Danger, GHS H Statement Toxic in contact with skin. Toxic if swallowed. Toxic if inhaled.

$576.53 - $576.53

Chemical Identifiers

CAS 14040-11-0
Molecular Formula C6O6W
Molecular Weight (g/mol) 351.91
MDL Number MFCD00011462
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221040500
SDS View Documents
Thermo Scientific Chemicals
221040500
50 g Glass bottle
Each for $576.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14040-11-0
C6O6W
351.91
MFCD00011462

Specifications

175.0°C
Yellow to Beige
Fine Crystalline Powder
99%
W(CO6)
06,658; 08,525; 05,142; 06,125; 07,71; 08,110; 13,51
351.91
99%
Glass bottle
150.0°C
14040-11-0
50 g
C6O6W
MFCD00011462
Solubility in water: insoluble. Other solubilities: soluble in organic solvents
351.91
Authentic
Tungsten hexacarbonyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comforta

GHS Signal Word: Danger

EINECSNumber : 237-880-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only