missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tungsten powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
$117.93 - $851.51
Chemical Identifiers
CAS | 7440-33-7 |
---|---|
Molecular Formula | W |
Molecular Weight (g/mol) | 183.84 |
MDL Number | MFCD00011461 |
InChI Key | WFKWXMTUELFFGS-UHFFFAOYSA-N |
Synonym | wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder |
PubChem CID | 23964 |
ChEBI | CHEBI:27998 |
IUPAC Name | tungsten |
SMILES | [W] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1040222
|
Thermo Scientific Chemicals
01040222 |
100 g |
Each for $117.93
|
|
|||||
AA1040236
|
Thermo Scientific Chemicals
01040236 |
500 g |
Each for $319.95
|
|
|||||
AA10402A3
|
Thermo Scientific Chemicals
010402A3 |
2 kg |
Each for $851.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-33-7 | |
183.84 | |
WFKWXMTUELFFGS-UHFFFAOYSA-N | |
23964 | |
tungsten |
W | |
MFCD00011461 | |
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
CHEBI:27998 | |
[W] |
Specifications
Gray | |
Light Gray powder | |
W | |
100 g | |
WFKWXMTUELFFGS-UHFFFAOYSA-N | |
tungsten | |
23964 | |
-100 Mesh | |
Tungsten |
7440-33-7 | |
99.9% (Metals basis) | |
MFCD00011461 | |
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
[W] | |
183.84 | |
CHEBI:27998 | |
Powder |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228-H500
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-143-9
RTECSNumber : YO7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only