missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tungsten powder, APS <1 micron, 99.95% (metals basis), Thermo Scientific Chemicals
$214.97 - $1530.32
Chemical Identifiers
CAS | 7440-33-7 |
---|---|
Molecular Formula | W |
Molecular Weight (g/mol) | 183.84 |
MDL Number | MFCD00011461 |
InChI Key | WFKWXMTUELFFGS-UHFFFAOYSA-N |
Synonym | wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder |
PubChem CID | 23964 |
ChEBI | CHEBI:27998 |
IUPAC Name | tungsten |
SMILES | [W] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4421022
|
Thermo Scientific Chemicals
04421022 |
100 g |
Each for $214.97
|
|
|||||
AA4421036
|
Thermo Scientific Chemicals
04421036 |
500 g |
Each for $588.25
|
|
|||||
AA44210A3
|
Thermo Scientific Chemicals
044210A3 |
2 kg |
Each for $1,530.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-33-7 | |
183.84 | |
WFKWXMTUELFFGS-UHFFFAOYSA-N | |
23964 | |
tungsten |
W | |
MFCD00011461 | |
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
CHEBI:27998 | |
[W] |
Specifications
Gray | |
W | |
UN3089 | |
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder | |
[W] | |
183.84 | |
CHEBI:27998 | |
APS <1 Micron | |
Tungsten powder |
7440-33-7 | |
MFCD00011461 | |
100 g | |
WFKWXMTUELFFGS-UHFFFAOYSA-N | |
tungsten | |
23964 | |
99.95% | |
Powder | |
(metals basis) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-143-9
RTECSNumber : YO7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only