Learn More
Tungsten(VI) ethoxide, Thermo Scientific Chemicals
Intermediate for sol-gel coatings
$130.59 - $497.85
Chemical Identifiers
CAS | 62571-53-3 |
---|---|
Molecular Formula | C12H30O6W |
Molecular Weight (g/mol) | 454.21 |
MDL Number | MFCD00156526 |
InChI Key | JHZFYDXXWXXVRM-UHFFFAOYSA-N |
Synonym | tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide |
PubChem CID | 13828235 |
IUPAC Name | ethanolate;tungsten |
SMILES | CCO[W](OCC)(OCC)(OCC)(OCC)OCC |
Description
Intermediate for sol-gel coatingsTungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. Also, it acts as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62571-53-3 | |
454.21 | |
JHZFYDXXWXXVRM-UHFFFAOYSA-N | |
13828235 | |
CCO[W](OCC)(OCC)(OCC)(OCC)OCC |
C12H30O6W | |
MFCD00156526 | |
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide | |
ethanolate;tungsten |
Specifications
62571-53-3 | |
White to Brown | |
W(OC2H5)6 | |
1 g | |
Moisture sensitive | |
Soluble in ethyl acetateSoluble in ethyl acetate,hydrocarbons and esters. | |
CCO[W](OCC)(OCC)(OCC)(OCC)OCC | |
454.21 | |
454.21 | |
Tungsten(VI) ethoxide |
150°C | |
C12H30O6W | |
MFCD00156526 | |
UN3181 | |
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide | |
JHZFYDXXWXXVRM-UHFFFAOYSA-N | |
ethanolate;tungsten | |
13828235 | |
Chunks and/or Powder |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only