missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tungsten wire basket; # turns, 9; ID(mm), 4; Ht(mm), 7; Wire leads(cm), 3.5; Wire dia(mm) 0.51, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 41177KK

 View more versions of this product

Catalog No. AA41177KK


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tungsten wire basket
7440-33-7
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Turns: 9, ID: (4mm), Ht(7mm), Wire leads (3.5cm), Wire dia (0.51mm)
W
10 Pc.
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

RTECSNumber : YO7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions