missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU packaged in an amber glass bottle.
Supplier: Ricca Chemical Company R88300004C
Description
Turbidity Standard; Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units); Ricca Chemical; 500 mL; Amber glass
- Turbidity Standard
- Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units)
- Amber glass
Specifications
Turbidity Standard | |
4000 NTU, Stock Formazin Suspension | |
C24H27NO5 | |
BCIPGSZQUDLGSY-UHFFFAOYNA-N | |
tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate | |
1.1 gal. | |
Amber Glass |
Turbidimeter | |
APHA/EPA for Turbidity, EP Primary Opalescent Suspension | |
9003-70-7 | |
CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O | |
409.48 | |
4 L |