missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turbidity Standard, 20 NTU, Polymer Bead Suspension, Ricca Chemical
Supplier: Ricca Chemical Company R88190001C
Specifications
| Turbidity Standard, 20 NTU, Polymer Bead Suspension | |
| 9003-53-6,26628-22-8,7732-18-5 | |
| 99.99,0.01,0.0009 | |
| Liquid | |
| MFCD00084450 | |
| 0°C | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| PPBRXRYQALVLMV-UHFFFAOYSA-N | |
| ethenylbenzene | |
| 1 L | |
| CHEBI:27452 |
| Amber Glass Bottle | |
| 99.99,0.01,0.0009 | |
| 1 | |
| (C8H8)n | |
| Colorless | |
| 100°C | |
| Miscible with water | |
| Analytical Standard | |
| *-CC(-*)C1=CC=CC=C1 | |
| 104.15 | |
| 7501 | |
| Laboratory |
Chemical Identifiers
| 9003-53-6 | |
| 104.15 | |
| PPBRXRYQALVLMV-UHFFFAOYSA-N | |
| CHEBI:27452 | |
| *-CC(-*)C1=CC=CC=C1 |
| (C8H8)n | |
| MFCD00084450 | |
| 7501 | |
| ethenylbenzene |
Safety and Handling
ShelfLife : 12 months
Recommended Storage : Ambient