Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU packaged in an amber glass bottle.
Supplier: Ricca Chemical Company 883016
Description
Turbidity Standard; Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units); Ricca Chemical; 500 mL; Amber glass
- Turbidity Standard
- Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units)
- Amber glass
Specifications
Turbidity Standard | |
16.9 oz. | |
Amber Glass |
Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units) | |
500 mL |