Impacto™ TURBOTOE™ Steel Safety Toe Cap
Impacto™ TURBOTOE Steel Safety Toe Cap protects the toe area from impact, crushing or stubbing in construction and heavy work environments that carries a risk of toe damage
Supplier:  Impacto™ TTUXXL
 View more versions of this product
Catalog No. 19-400-035
Explore available promotions
Description
Description
Impacto™ Turbotoe steel toe cap overshoes
  • Made of flexible waterproof that do not harden or crack
  • Treads under sole help prevent slipping
  • For visitors, casual or temporary workers
  • Fits tightly over existing shoes, providing a solid protecting cap
  • Comfortably worn over low heel, dress, running shoes and most boots
  • Color-coded for sizing
  • Meet ASTM F2412/F2413 impact and compression standard
Specifications
Specifications
CE
PVC
Steel
Yes
Green
2X to Large, Men: 13 to 14
Unisex
Turbotoe Safety Toe Cap