TWD Scientific™ Tradewinds Direct Weighing Funnels
Disposable polypropylene funnels in four different sizes
Supplier: TWD Scientific™ DPWFPP3L
Description
Disposable polypropylene funnels in four different sizes
- Designed to lie flat on a weighing balance Place funnel directly on balance, weigh out needed solids or liquids, transfer sample into flask
- Pouring necks designed to easily fit in volumetric flasks and test tubes
Specifications
20 mL | |
88 mm | |
Serve as weighing dishes plus sample transfer funnels | |
Tradewinds Direct Weighing Funnel |
Polypropylene | |
Size: Large | |
500mL and 1L volumetric flasks |