Thermo Scientific™ TX-150 Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Perform a a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor, which is highly configurable and convenient.
Supplier:  Thermo Scientific™ 75005701
Catalog No. 75-005-701
Explore available promotions
Description
Description
Perform a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor. This rotor accommodates up to 24 x 5/7mL blood collection tubes or 8 x 50mL conical tubes at 4,500rpm/ 3,260 x g, and provides additional flexibility for swing-out processing needs with available adapters. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ ClickSeal™ biocontainment lids—shorten run set-up time while providing peace-of-mind that samples are secure.
Designed for a broad range of swing-out processing needs at high speed and high capacity with dedicated buckets available for 8 conical tubes capacity or round buckets with ClickSeal biocontainment lids certified by Public Health England, Porton Down, UK.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
1. No tools are required
2. No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Round buckets (75005702), 50mL Conical buckets (75005703) ClickSeal biocontainment lids (75005707) and replacement O-rings (75005724) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Specifications
Specifications
TX-150 Swinging Bucket Rotor
4 x 145 mL
4,500 rpm
12968
50 mm
90°
Swinging Bucket Rotor
3,260 x g
144 mm
100 mm
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Yes
Videos
SDS
Documents
Documents
Accessories