Thermo Scientific™ TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
$502.62 - $566.25
Products 12
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
75-003-771
View Documents
Thermo Scientific™
75003771
15mL Conical tube
Each for $541.04
75-003-785
View Documents
Thermo Scientific™
75003785
5/7mL Blood Collection tube
Each for $509.83
75-003-801
View Documents
Thermo Scientific™
75003801
100mL Round-bottom tube
Each for $502.62
75-003-802
View Documents
Thermo Scientific™
75003802
50mL DIN Round-bottom tube
Each for $502.62
75-003-803
View Documents
Thermo Scientific™
75003803
50mL Conical tube
Each for $541.04
75-003-804
View Documents
Thermo Scientific™
75003804
25mL Universal tube
Each for $504.50
75-003-805
View Documents
Thermo Scientific™
75003805
25mL DIN Round-/Flat-bottom tube
Each for $502.62
75-003-807
View Documents
Thermo Scientific™
75003807
14mL Conical Urine tube
Each for $528.04
75-003-808
View Documents
Thermo Scientific™
75003808
10mL Blood Collection or 15mL Corex /Kimble tube
Each for $541.04
75-003-809
View Documents
Thermo Scientific™
75003809
15mL Blood Collection tube
Each for $502.62
75-003-810
View Documents
Thermo Scientific™
75003810
5/7mL Round-bottom tube
Each for $528.04
75-003-812
View Documents
Thermo Scientific™
75003812
1.5/2.0mL Conical Microtube
Each for $566.25
Description
Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
SDS
missing translation for 'documents'
missing translation for 'documents'