Learn More
DuPont™ Tychem™ 10,000 Class 2, Level A Suits
Extremely durable to abrasion, tearing, punctures, and cuts
Supplier: DuPont™ TK612TLYLG000100
Description
Tychem TK Class 2 Level A Suits—Use NFPA 1994 Class 2 suits in situations where an unidentified chemical/biological substance of unknown concentration poses an immediate threat.
- Extra-wide three-layer faceshield
- Double taped seams
- Gastight zipper closure
- Double storm flaps
- Attached three-layer gloves and sock boots
- High-visibility lime yellow
- Gastight zipper lock closure with double storm flaps
- Double-taped seams
- Two-layer 40mil PVC faceshield
- Three exhaust valves
- Attached three-layer glove system
- Attached sock boots with outer boot flaps
Compliance
Meets NFPA 1994 Standard on Protective Ensembles for First Responders to Hazardous Materials Emergencies and CBRN Terorrism Incident, Class 2 requirements.
NOTE: Product safety information supplied corresponds to current knowledge on the subject. This information may be subject to revisions as new knowledge and experience becomes available. DuPont makes no warranties and assumes no liability in connection with any use of this information. It is the end user's responsibility to determine level of risk and select proper protective equipment for end-use conditions. Nothing in this product offering is to be considered license to operate under or a recommendation to infringe upon any patent rights.
Specifications
NFPA 1994, Class 2 | |
Industrial, HazMat and domestic preparedness applications | |
Large | |
Expanded faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal multilayer laminate/external Neoprene size 10 gloves, socks with outer boot flaps, soles; Specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals, excellent chemical barrier properties and offers an extremely durable Fabric that is puncture and tear-resistant | |
1.79 to 1.92 m | |
29.25 in. | |
74.29 cm | |
46.75 to 50.25 in. | |
Front Entry |
Lime Yellow | |
Tychem | |
Front zipper | |
5.9 to 6.3 ft. | |
30.75 in. | |
78.105 cm | |
Taped | |
118.75 to 127.65 cm | |
27 mil |