DuPont™ Tychem™ 10,000 Level A Suits
Supplier:  DuPont™ TK555TLY2X000100
 View more versions of this product
Catalog No. 19-715-234
Explore available promotions
Description
Description
Fully Encapsulated Level A Suits are ideal for industrial, HazMat and domestic preparedness situations. High-visibility lime yellow.
  • Double taped seams
  • Three-layer faceshield (PVC 40mil/PTFE 5mil/PVC 20mil)
  • Gas-tight PVC zipper closure
  • Double storm flaps with Velcro™ closure
  • Attached internal Barrier gloves, outer butyl gloves and sock boots with outer boot flaps
  • Knee wear pads
  • Two exhaust valves
  • Internal adjustment belt
  • Garments successfully tested against more than 260 challenge chemicals
Compliance
Meets OSHA 1910.132 (a) standards.
Specifications
Specifications
Lime Yellow
Tychem
Gas-tight zipper and double storm flaps with hook-and-loop closure
6.3 to 6.5 ft.
32.5 in.
82.55 cm
Butyl gloves
49.75 to 53.25 in.
Rear entry
Industrial, HazMat and domestic preparedness applications
2X-Large
Expanded faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal multilayer laminate/external Butyl size 10 gloves, socks with outer boot flaps, soles; Specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals, excellent chemical barrier properties and offers an extremely durable Fabric that is puncture and tear-resistant
1.92 to 1.98 m
31 in.
78.74 cm
Taped
126.36 to 135.25 cm
27 mil